Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) bromide, 98%, Thermo Scientific Chemicals

Catalog No. p-4904558
Click to view available options
Quantity:
10 g

Titanium(IV) bromide is used as a Lewis acid catalyst in organic synthesis. It reacts with tetrahydrofurnan and 2-methylpyridine and forms adducts such as TiBr4(THF)2 and TiBr4(2-MePy). It is also used for the formation of titanium silicide thin films on silicon substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-68-6
Molecular Formula TiBr4
MDL Number MFCD00011265

Specifications

Boiling Point 233°C
Melting Point 39°C
Color Orange
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 98%
Solubility Information Soluble in absolute alcohol. Decomposes in waterSoluble in chlorocarbons,benzene and alcohol.
Formula Weight 367.54
Odor Acrid
Sensitivity Moisture sensitive
Density 2.6
Chemical Name or Material Titanium(IV) bromide
Show More Show Less
Hazard Category H314
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-185-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.