Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) chloride, 0.09M solution in 20% HCl, AcroSeal™

Catalog No. ac428351000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Titanium(IV) chloride
Color Colorless to Yellow
CAS 7550-45-0, 7647-01-0, 7732-18-5
Physical Form Solution
Quantity 100 mL
Molecular Formula Cl4Ti
MDL Number MFCD00011267
Merck Index 15, 9634
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
Solubility Information Solubility in water: soluble
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
IUPAC Name titanium(4+);tetrachloride
Molecular Weight (g/mol) 189.71
PubChem CID 160960
Formula Weight 189.71
Concentration or Composition (by Analyte or Components) 0.08 to 0.10M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.099
Density 1.0990g/mL
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.