Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) chloride, 99.0% min, typically 99.6%

Catalog No. p-4408025
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L

Catalog No. AA14713AE

Supplier: Thermo Scientific Chemicals 014713AE

Only null left
Add to Cart

For Use in Manufacture of Titanium Pigments and Metal, As a Catalyst in organic Syntheses, in Iridescent Glass | CAS: 7550-45-0 | Cl4Ti | 189.667 g/mol

In manufacture of titanium pigments and metal, as a catalyst in organic syntheses, in iridescent glass, as a smoke screenTitanium(IV) chloride is used to make titanium metal, pigment titanium dioxide and is used in smoke screens. It is involved in the activation of pyrrolidines for better conversion through a modified Bouveault reaction, to the corresponding alpha, alpha -dimethylamines. In organometllic chemistry, it is used to prepare complexes like titanocene dichloride, which is a precursor to Ziegler-Natta catalysts. Tebbe’s reagent is an aluminum containing derivative of titanocene used for the olefination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7550-45-0
Molecular Formula Cl4Ti
Molecular Weight (g/mol) 189.667
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]

Specifications

Boiling Point 136°C to 137°C
Melting Point 25°C
Color Yellow
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 99.0% min., typically 99.6%
Solubility Information Soluble in dilute hydrochloric acid,alcohol and alkalies.Miscible with ethanol,hydrochloric acid and organic solvents.
Formula Weight 189.69
Refractive Index 1.61
Sensitivity Moisture sensitive
Appearance Colorless to yellow liquid
Density 1.726 g/cm3
Chemical Name or Material Titanium(IV) chloride
Show More Show Less
Hazard Category H314-H330-H335
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 6.1, Packing Group: I
EINECSNumber 231-441-9
RTECSNumber XR1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.