Alfa Aesar™ Titanium(IV) ethoxide, TiO2 33% min

Manufacturer: Alfa Aesar™ 7714206

Quantity 5g

 View more versions of this product

Catalog No. AA7714206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3087-36-3
    Molecular Formula C8H20O4Ti
    Linear Formula Ti(OCH2CH3)4
    Formula Weight 228.15
    MDL Number MFCD00009071
    UN Number UN2924
    Synonym Tetraethyl titanate
    Chemical Name or Material Titanium (IV) ethoxide

    Specifications

    Formula Weight 228.15
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00009071
    Flash Point 28°C (84°F)
    Linear Formula Ti(OCH2CH3)4
    CAS 3087-36-3
    UN Number UN2924
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H226-H319
    Flammable liquid and vapor.
    Causes serious eye irritation.
    Density 1.088g/mL
    TSCA TSCA
    Odor Alcohol-like
    Synonym Tetraethyl titanate
    Color Pale yellow
    Melting Point −40°C
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Physical Form Liquid
    Sensitivity Moisture Sensitive
    Boiling Point 150° to 152°C (10mmHg)
    Concentration TiO2 33% min
    Refractive Index 1.5043
    Molecular Formula C8H20O4Ti
    Solubility Information Soluble in organic solvents; Decomposes in water
    Chemical Name or Material Titanium (IV) ethoxide

    GHS H Statement
    H226-H319
    Flammable liquid and vapor.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit