Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) ethoxide, TiO{2} 33% min

Catalog No. p-7080340
Click to view available options
Quantity:
5 g
50 g
250 g

CAS: 3087-36-3 | C8H20O4Ti | 228.111 g/mol

Titanium(IV) ethoxide is used in organic synthesis and material science. It is involved in the synthesis of anti-perspirants and antifungal agents. Furthermore, it is used to prepare carbon films in association with silicon dioxide. It is also used as a precursor for the synthesis of Ti4+ ions titanium nanoparticles by various methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3087-36-3
Molecular Formula C8H20O4Ti
Molecular Weight (g/mol) 228.111
MDL Number MFCD00009071
InChI Key JMXKSZRRTHPKDL-UHFFFAOYSA-N
Synonym titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
PubChem CID 76524
IUPAC Name ethanolate;titanium(4+)
SMILES CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

Specifications

Boiling Point 150°C to 152°C (10mmHg)
Melting Point -40°C
Color Yellow
Physical Form Liquid
Quantity 5 g
Linear Formula Ti(OCH2CH3)4
UN Number UN2924
Flash Point 28°C (84°F)
Solubility Information Soluble in organic solvents; Decomposes in water.
Formula Weight 228.15
Concentration or Composition (by Analyte or Components) TiO2 33% min
Odor Alcohol-like
Refractive Index 1.5043
Sensitivity Moisture sensitive
Density 1.088 g/mL
Chemical Name or Material Titanium (IV) ethoxide
Show More Show Less
Hazard Category H226-H319-H336
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 221-410-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.