Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) iodide, anhydrous, 99.9% (metals basis)

Catalog No. p-7051172
Click to view available options
Quantity:
10 g
50 g

CAS: 7720-83-4

Titanium(IV) iodide, anhydrous is used in the CVD of titanium nitride films with optimized characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7720-83-4
MDL Number MFCD00043077

Specifications

Melting Point 150°C
Physical Form Lump
Quantity 10 g
UN Number UN3260
Solubility Information Soluble in dichloromethane,chloroform,carbon disulfide.
Formula Weight 555.52
Percent Purity 99.9%
Odor Acrid
Sensitivity Moisture sensitive
Density 4.30 g/mL
Assay (metals basis)
Chemical Name or Material Titanium(IV) iodide, Anhydrous
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-754-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.