Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) isopropoxide, 97+%

Catalog No. AA7711557
Click to view available options
Quantity:
10 g
100 g
500 g
4 x 500 g

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids | CAS: 546-68-9 | C12H28O4Ti | 284.219 g/mol

Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids.Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-68-9
Molecular Formula C12H28O4Ti
Molecular Weight (g/mol) 284.219
MDL Number MFCD00008871
InChI Key VXUYXOFXAQZZMF-UHFFFAOYSA-N
Synonym titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
PubChem CID 11026
IUPAC Name propan-2-olate;titanium(4+)
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Specifications

Boiling Point 232°C
Melting Point 16°C to 20°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 4 x 500 g
Linear Formula Ti[OCH(CH3)2]4
UN Number UN2413
Beilstein 3679474
Flash Point 46°C (115°F)
Merck Index 14,9480
Solubility Information Soluble in anhydrous ethanol, ether, benzene and chloroform; Decomposes in water
Formula Weight 284.23
Percent Purity ≥97%
Refractive Index 1.464
Sensitivity Moisture sensitive
Density 0.955 g/mL
Chemical Name or Material Titanium (IV) isopropoxide
Show More Show Less
Hazard Category H226-H319-H331-H335-H336
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE
EINECSNumber 208-909-6
RTECSNumber NT8060000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.