Learn More
Titanium(IV) n-butoxide, 99+%
Description
Titanium(IV) n-butoxide is used in the preparation of nanosized titania powders in the anatase form and ferroelectric bismuth titanate thin films. It is also used as intermediates, paint additives, coating additives, processing aids and as process regulators. It is involved in the preparation of nanocrystalline titanium dioxide powders at room temperature.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 310°C to 314°C |
Melting Point | -55°C |
Color | Yellow |
Physical Form | Liquid |
Quantity | 500 g |
Linear Formula | Ti[O(CH2)3CH3]4 |
UN Number | UN2924 |
Beilstein | 4148236 |
Flash Point | 41°C (170°F) |
Solubility Information | Miscible with aliphatic,aromatic,chlorinated and oxygenated solvents. |
Show More |
Safety and Handling
Hazard Category | H226-H315-H318-H335-H336 |
Hazard Statement | GHS H Statement H314-H318-H226 Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
EINECSNumber | 227-006-8 |
RTECSNumber | XR1585000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.