Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) n-propoxide, VERTEC™ NPT, 98+%, Thermo Scientific™

Catalog No. AA4467322
Click to view available options
Quantity:
25 g
100 g
500 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3087-37-4
Molecular Formula C3H8OTi+4
Molecular Weight (g/mol) 107.963
MDL Number MFCD00009365
InChI Key ITYITLIANRYTCY-UHFFFAOYSA-N
Synonym titanium propoxide, titanium 4+ ion propanol
PubChem CID 91996326
IUPAC Name propan-1-ol;titanium(4+)
SMILES CCCO.[Ti+4]

Specifications

Boiling Point 58°C (1mmHg)
Melting Point 20°C
Physical Form Liquid
Quantity 100 g
UN Number UN2413
Flash Point 42°C (109°F)
Formula Weight 284.25
Percent Purity ≥98%
Refractive Index 1.4986
Density 0.955g/mL
Chemical Name or Material Titanium (IV) n-propoxide, VERTEC™ NPT
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.