Learn More
Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion
Description
UV protection in clear coatings. Photo electrodes of varying properties can be made using colloidal suspensions of titanium(IV) oxide nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 368°C |
| Quantity | 1 kg |
| Solubility Information | Insoluble in water. |
| Formula Weight | 79.9 |
| Concentration or Composition (by Analyte or Components) | 20 to 35% in H2O |
| Density | 1.26 g/mL |
| Chemical Name or Material | Titanium (IV) oxide, Colloidal dispersion |
Safety and Handling
| Hazard Category | H351 |
| Hazard Statement | GHS H Statement H351 Suspected of causing cancer. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.