Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) oxide, Puratronic™, 99.995% (metals basis)

Catalog No. aa1089709
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 13463-67-7 | O2Ti | 79.87 g/mol

Titanium(IV) oxide is used in paints, varnishes, paper, plastics printing inks, sun screen, fibers, rubber, cosmetic products, anti-fogging coatings, foodstuffs, pure titanium, glass, glass ceramics, memristor, electrical ceramics, catalysts, electric conductors and chemical intermediates. It exhibit excellent photocatalytic properties (Honda-Fujishima effect) In addition, titanium dioxide is also used to cleave protein at the site where proline is present.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Boiling Point 2500°C to 3000°C
Melting Point 1830°C to 1850°C
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.995% (metals basis)
Solubility Information Slightly soluble in hydrofluoric acid and hot concentrated sulfuric acid. Insoluble in water,hydrochloric acid,dilute sulfuric acid,and organic solvents.
Formula Weight 79.9
Density 4.26
Chemical Name or Material Titanium(IV) oxide
Hazard Category H351
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 236-675-5
RTECSNumber XR2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.