Learn More
Titanium(IV) oxide, Puratronic™, 99.995% (metals basis)
Description
Titanium(IV) oxide is used in paints, varnishes, paper, plastics printing inks, sun screen, fibers, rubber, cosmetic products, anti-fogging coatings, foodstuffs, pure titanium, glass, glass ceramics, memristor, electrical ceramics, catalysts, electric conductors and chemical intermediates. It exhibit excellent photocatalytic properties (Honda-Fujishima effect) In addition, titanium dioxide is also used to cleave protein at the site where proline is present.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 2500°C to 3000°C |
Melting Point | 1830°C to 1850°C |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.995% (metals basis) |
Solubility Information | Slightly soluble in hydrofluoric acid and hot concentrated sulfuric acid. Insoluble in water,hydrochloric acid,dilute sulfuric acid,and organic solvents. |
Formula Weight | 79.9 |
Density | 4.26 |
Chemical Name or Material | Titanium(IV) oxide |
Safety and Handling
Hazard Category | H351 |
Precautionary Statement | P201-P202-P281-P308+P313-P501c |
EINECSNumber | 236-675-5 |
RTECSNumber | XR2275000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.