Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) oxide, 99.999%, (trace metal basis), predominantly rutile, Thermo Scientific Chemicals

Catalog No. AC194340010
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Boiling Point 2900.0°C
Melting Point 1855.0°C
Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.999%
Linear Formula TiO2
Merck Index 15,9628
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
Formula Weight 79.88
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.23
Density 4.2300g/mL
Chemical Name or Material Titanium(IV) oxide, 99.999%
Show More Show Less
EINECSNumber 236-675-5
RTECSNumber XR2275000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.