Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) oxide, anatase, nanopowder, 99.7% (metals basis)

Catalog No. p-4408030
Click to view available options
Quantity:
500 g

CAS: 1317-70-0 | O2Ti | 79.87 g/mol

Titanium(IV) oxide is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-70-0
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269,MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Physical Form Powder
Quantity 500 g
Assay Percent Range 99.7% (metals basis)
Solubility Information Insoluble in water,dilute acids,organic acids,and dilute alkalis.
Formula Weight 79.9
Chemical Name or Material Titanium(IV) oxide
What is the difference between Titanium (IV) oxide, rutile and Titanium (IV) oxide, anatase?

Rutile and anatase are polymorphs of TiO2, which are both naturally occurring. Rutile is a source for titanium smelting and is used in the semiconductor industry to produce dye-based solar cells (Grätzel cells). Anatase shows a high photocatalytic activity, which limits the usability as a pigment in polymers, but also opens certain applications as a catalyst. Anatase has a higher photocatalytic activity than rutile but is thermodynamically less stable.

EINECSNumber 215-280-1
RTECSNumber BV6550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.