Learn More
Titanium(IV) sulfide, 99.8% (metals basis)
Description
Titanium(IV) sulfide is used in solid lubricant, cathode material in rechargeable batteries, corrosion resistance alloys, solid-state lithium batteries, hybrid electric vehicles and plug-in electric vehicles. It is used in the synthesis of nanoclusters, nanodisks and nanotubes. It has high electrical conductivity. Titanium electrodes are used in chloralkali electrolysis, electrodialysis, galvanic technology, light bulbs, and bone pins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.8%, metal basis |
Solubility Information | Titanium(IV) sulfide is insoluble in water and moderately acid soluble. |
IUPAC Name | bis(sulfanylidene)titanium |
Formula Weight | 112.03 |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Titanium(IV) Sulfide |
Safety and Handling
Hazard Category | H252-H315-H319-H335 |
Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. |
Precautionary Statement | P235+P410-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
DOTInformation | DOT Class: 4.2; Packaging Group: III |
EINECSNumber | 234-883-0 |
RTECSNumber | XR2460000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.