Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium(IV) tert-butoxide, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3087-39-6
C16H36O4Ti
340.33
MFCD00040554
GRWPYGBKJYICOO-UHFFFAOYSA-N
unii-px6yr1s99s,px6yr1s99s,titanium iv tert-butoxide,titanium tetra-tert-butoxide,titanium tert-butoxide,titanum tetra t-butoxide,tetra-tert-butoxytitanium iv,titanium 4+ 2-methylpropan-2-olate
6451515
2-methylpropan-2-olate;titanium(4+)
CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].[Ti+4]
3087-39-6
C16H36O4Ti
340.33
MFCD00040554
GRWPYGBKJYICOO-UHFFFAOYSA-N
unii-px6yr1s99s,px6yr1s99s,titanium iv tert-butoxide,titanium tetra-tert-butoxide,titanium tert-butoxide,titanum tetra t-butoxide,tetra-tert-butoxytitanium iv,titanium 4+ 2-methylpropan-2-olate
6451515
2-methylpropan-2-olate;titanium(4+)
CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].[Ti+4]
Lewis acid
Color Colorless
Density 0.8893g/mL
Quantity 5g
Boiling Point 160.0°C to 165.0°C (15.0mmHg)
Specific Gravity 0.8893
Packaging Glass bottle
Flash Point 47°C
Assay Percent Range 98%
Linear Formula Ti[OC(CH3)3]4
Beilstein 01, IV, 1613
Formula Weight 340.33
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Titanium(IV) tert-butoxide
Show More Show Less

GHS H Statement:
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning