Learn More
Titanium nitride, 99.7% (metals basis)
Description
Titanium nitride is a hard ceramic material used as a coating on titanium alloys, steel, carbide and aluminum components. It finds application in jewelry and high-performance ceramic materials due to its gold colored surface and in medical devices such as scalpel blades and orthopedic bone saw blades. Further, it is used as a conductive barrier in microelectronics and electrodes in bioelectronic applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2950°C |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.7% (metal basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 61.91 |
Density | 5.22 |
Chemical Name or Material | Titanium nitride |
Safety and Handling
EINECSNumber | 247-117-5 |
RTECSNumber | XR2230000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.