Alfa Aesar™ Titanium, plasma standard solution, Specpure™, Ti 1000µg/mL

Manufacturer: Alfa Aesar™ 38720AD

Quantity 50mL

 View more versions of this product

Catalog No. AA38720AD

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00011264
    Chemical Name or Material Titanium plasma standard solution, Ti 1000μg/mL
    Grade Specpure™

    Specifications

    Quantity 50mL
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00011264
    Chemical Name or Material Titanium plasma standard solution, Ti 1000μg/mL
    Grade Specpure™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit