Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Titanium silicide, 99.5% (metals basis)
Click to view available options
Quantity:
50 g
250 g
Description
It is used as source, gate and drain contacts and local interconnections in CMOS integrated circuits. it is used in the C54 phase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2130°C |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 99.5%, metal basis |
| Solubility Information | Soluble in hydrofluoric acid. Insoluble in water. |
| Formula Weight | 323.76 |
| Chemical Name or Material | Titanium Silicide |
Safety and Handling
| Hazard Category | H228-H315-H319-H335 |
| Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | DOT Class: 4.1; Packaging Group: III |
| EINECSNumber | 235-082-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction