Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Titanium Standard, 1mL = 1mg Ti, 1000ppm Ti, Ricca Chemical
SDP

Catalog No. 860016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Glass amber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
860016 500 mL Glass amber
8600-4 120 mL Glass amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 860016 Supplier Ricca Chemical Company Supplier No. 860016

Chemical Identifiers

CAS 7440-32-6 , 7647-01-0 , 7732-18-5
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Packaging Glass amber
Concentration or Composition Notes 1,000ppm Ti
Equivalence 1 mL = 1 mg Ti
Physical Form Liquid
Color Colorless to Purple
Quantity 500 mL
Chemical Name or Material Titanium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.