Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Titin Monoclonal Antibody (T11)
GREENER_CHOICE

Catalog No. MA122379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA122379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA122379 Supplier Invitrogen™ Supplier No. MA122379
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: skeletal muscle.

Monoclonal Anti-Titin can be used for study of the elastic filaments within sarcomeric structures. It is also useful as a differentiation marker in the separation of rhabdomyosarcomas from other muscle tumors. By indirect immunofluorescence the antibody displays a typical striated staining pattern on frozen sections of chicken skeletal and cardiac muscle tissues. Stains the region of the A-I junction by indirect immunofluorescence. It shows a decoration line 0.05 mm from the end of the A band in electron micro-graphs. In immunoblotting using total extracts of chicken breast muscle the antibody reacts specifically with both bands of the titin doublet, and showed no reaction with nebulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Titin
Applications Immunohistochemistry (Frozen), Immunomicroscopy, Western Blot
Classification Monoclonal
Clone T11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 15mM sodium azide
Gene TTN
Gene Accession No. 0
Gene Alias 1100001C23Rik; 2.7.11.1; 2310036G12Rik; 2310057K23Rik; 2310074I15Rik; AF006999; AV006427; CMD1G; CMH9; CMPD4; Connectin; connectin/titin; D330041I19Rik; D830007G01Rik; DKFZp451N061; EOMFC; FLJ26020; FLJ26409; FLJ32040; FLJ34413; FLJ39564; FLJ43066; HMERF; L56; LGMD2J; mdm; MYLK5; rhabdomyosarcoma antigen MU-RMS-40.14; shru; structural muscle protein titin; titin; titin protein homolog; titin sequence3; TMD; TTN
Gene Symbols TTN
Host Species Mouse
Immunogen Titin/nebulin fraction from chicken breast muscle.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 424126
Target Species Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.