Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Titin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583120 Supplier Invitrogen™ Supplier No. PA583120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HFLSILTIDT SDAEDYSCVL VEDENVKTTA KLIVEGAVVE FVKELQDIEV PESYSGELEC IVSPENIEGK WYHNDVELKS NGKYTITSRR GRQNLTVKDV TKEDQGEYSF VIDGKKTTCK LKMK Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 30%.

Monoclonal Anti-Titin can be used for study of the elastic filaments within sarcomeric structures. It is also useful as a differentiation marker in the separation of rhabdomyosarcomas from other muscle tumors. By indirect immunofluorescence the antibody displays a typical striated staining pattern on frozen sections of chicken skeletal and cardiac muscle tissues. Stains the region of the A-I junction by indirect immunofluorescence. It shows a decoration line 0.05 mm from the end of the A band in electron micro-graphs. In immunoblotting using total extracts of chicken breast muscle the antibody reacts specifically with both bands of the titin doublet, and showed no reaction with nebulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Titin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTN
Gene Accession No. Q8WZ42
Gene Alias 1100001C23Rik; 2.7.11.1; 2310036G12Rik; 2310057K23Rik; 2310074I15Rik; AF006999; AV006427; CMD1G; CMH9; CMPD4; Connectin; connectin/titin; D330041I19Rik; D830007G01Rik; DKFZp451N061; EOMFC; FLJ26020; FLJ26409; FLJ32040; FLJ34413; FLJ39564; FLJ43066; HMERF; L56; LGMD2J; mdm; MYLK5; rhabdomyosarcoma antigen MU-RMS-40.14; shru; structural muscle protein titin; titin; titin protein homolog; titin sequence3; TMD; TTN
Gene Symbols TTN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTN. Recombinant protein control fragment (Product #RP-108190).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7273
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.