Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Titration Solvent, ASTM D664, D974, D3242, Reagents
Titration Solvent, ASTM D664, D974, D3242, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CT1250004E4
Specifications
| Titration Solvent, ASTM D664, D974, D3242 | |
| 108-88-3, 67-63-0, 7732-18-5 | |
| Reagent | |
| 0.8245 | |
| Non-sterile |
| Colorless | |
| 90°C | |
| Tin Can | |
| 4 L | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction