Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TJAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542266
Description
Peptide sequence: MTSAAPAKKP YRKAPPEHRE LRLEIPGSRL EQEEPLTDAE RMKLLQEENE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
This gene encodes a tight junction-associated protein. Incorporation of the encoded protein into tight junctions occurs at a late stage of formation of the junctions. The encoded protein localizes to the Golgi and may function in vesicle trafficking. Alternatively spliced transcript variants have been described. A related pseudogene exists on the X chromosome.
Specifications
TJAP1 | |
Polyclonal | |
Unconjugated | |
TJAP1 | |
0610041D19Rik; AI415281; AW121008; hdlg-binding protein Pilt; Pilt; Protein incorporated later into tight junctions; sb:eu465; si:dkey-109m21.1; tight junction associated protein 1; tight junction associated protein 1 (peripheral); tight junction associated protein 1 (peripheral) isoform 1; tight junction associated protein 1 (peripheral) isoform 2; Tight junction protein 4; tight junction protein 4 (peripheral); tight junction-associated protein 1; TJAP1; TJP4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
93643 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5JTD0 | |
TJAP1 | |
Synthetic peptide directed towards the N-terminal of human TJAP1 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction