Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TJAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556376 Supplier Invitrogen™ Supplier No. PA556376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSPEEELPLP AFEKLNPYPT PSPPHPLYPG RRVIEFSEDK VRIPRNSPLP NCTYATRQAI SLSLVEEGSE RARPSPVPST PASAQASPHH QPSPAPLTLS AP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 83%.

This gene encodes a tight junction-associated protein. Incorporation of the encoded protein into tight junctions occurs at a late stage of formation of the junctions. The encoded protein localizes to the Golgi and may function in vesicle trafficking. Alternatively spliced transcript variants have been described. A related pseudogene exists on the X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TJAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TJAP1
Gene Accession No. Q5JTD0
Gene Alias 0610041D19Rik; AI415281; AW121008; hdlg-binding protein Pilt; Pilt; Protein incorporated later into tight junctions; sb:eu465; si:dkey-109m21.1; tight junction associated protein 1; tight junction associated protein 1 (peripheral); tight junction associated protein 1 (peripheral) isoform 1; tight junction associated protein 1 (peripheral) isoform 2; Tight junction protein 4; tight junction protein 4 (peripheral); tight junction-associated protein 1; TJAP1; TJP4
Gene Symbols TJAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TJAP1. Recombinant protein control fragment (Product #RP-94866).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93643
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.