Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TJP1 Mouse anti-Human, Clone: ZO1-1A12 (FITC), Abnova™

Catalog No. 89035590 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-590 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-590 Supplier Abnova Corporation Supplier No. MAB14193

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human TJP1.

  • High specificity
  • High reactivity

Specifications

Antigen TJP1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone ZO1-1A12
Conjugate FITC
Description Mouse monoclonal antibody raised against partial recombinant human TJP1.
Dilution ELISA (0.1-1 ug/mL) Immunocytochemistry (1-10 ug/mL) Immunofluorescence (1-10 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 1% BSA, 0.09% sodium azide).
Gene Accession No. Q07157
Gene Alias DKFZp686M05161/MGC133289/ZO-1
Gene Symbols TJP1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 334-634 of human TJP1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7082
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.