Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TJP3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
| Antigen | TJP3 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TJP3 Polyclonal specifically detects TJP3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| TJP3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| MGC119546, Tight junction protein 3, tight junction protein 3 (zona occludens 3), ZO-3, ZO3tight junction protein ZO-3, zona occludens 3, Zona occludens protein 3, Zonula occludens protein 3 | |
| TJP3 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 27134 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PYTDVDDEPPAPALARSSEPVQADESQSPRDRGRISAHQGAQVDSRHPQGQWRQDSMRTYEREALKKKFTRVHDAESSDEDGYDWGP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title