Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TK1 Monoclonal Antibody (C3)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | TK1 |
---|---|
Clone | C3 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541680
|
Invitrogen™
MA541680 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543560
|
Invitrogen™
MA543560 |
20 μL |
Each for $243.00
|
|
|||||
Description
TK1 Monoclonal Antibody for Western Blot, ICC/IF
Thymidine Kinase (TK1) is a highly conserved phosphotransferase that is present in most living cells. Thymidine Kinase catalyzes the phosphorylation reaction: deoxythymidine + ATP = deoxythymidine 5'-phosphate + ADP; it is thus involved in the reaction chain to introduce deoxythymidine into the DNA. Thymidine kinase is required for the action of many antiviral drugs, such as azidothymidine (AZT), and is also used to select hybridoma cell lines in the production of monoclonal antibodies. Thymidine Kinase has many clinical applications as it is only present in anticipation of cell division.Specifications
TK1 | |
1 mg/mL | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P04183 | |
7083 | |
Recombinant protein Thymidine Kinase 1, Soluble. The antigen corresponds to amino acid range 2-234 of the target protein. | |
Antibody |
C3 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
TK1 | |
D530002A18Rik; EC 2.7.1.21; KITH; thymidine kinase 1; thymidine kinase 1 soluble isoform; thymidine kinase 1, soluble; thymidine kinase, cytosolic; thymidine kinase-1; Tk; Tk1; Tk-1; Tk1a; Tk1b; TK2 | |
TK1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title