Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598195
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598195 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598195 Supplier Invitrogen™ Supplier No. PA598195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TK1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Thymidine Kinase (TK1) is a highly conserved phosphotransferase that is present in most living cells. Thymidine Kinase catalyzes the phosphorylation reaction: deoxythymidine + ATP = deoxythymidine 5'-phosphate + ADP; it is thus involved in the reaction chain to introduce deoxythymidine into the DNA. Thymidine kinase is required for the action of many antiviral drugs, such as azidothymidine (AZT), and is also used to select hybridoma cell lines in the production of monoclonal antibodies. Thymidine Kinase has many clinical applications as it is only present in anticipation of cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TK1
Gene Accession No. P04183
Gene Alias D530002A18Rik; EC 2.7.1.21; KITH; thymidine kinase 1; thymidine kinase 1 soluble isoform; thymidine kinase 1, soluble; thymidine kinase, cytosolic; thymidine kinase-1; Tk; Tk1; Tk-1; Tk1a; Tk1b; TK2
Gene Symbols TK1
Host Species Rabbit
Immunogen Recombinant Human Thymidine kinase, cytosolic protein (2-234AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7083
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.