Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TK15 Epitope Tag Antibody (TK 15), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250335H
Description
TK15 Epitope Tag Monoclonal antibody specifically detects TK15 Epitope Tag in Epitope Tag samples. It is validated for Western Blot.Specifications
TK15 Epitope Tag | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Recombinant full length Xenopus Orc1p protein | |
0.1 mL | |
Primary | |
Purified | |
Epitope Tag |
Western Blot | |
TK 15 | |
Western Blot | |
Mouse | |
Protein A purified | |
RUO | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction