Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TKS5, Mouse anti-Human, Clone: 13H6.3, Millipore Sigma™

Catalog No. MABT33625UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT33625UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT33625UG Supplier MilliporeSigma Supplier No. MABT33625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TKS5, clone 13H6.3, Cat. No. MABT336, ia a mouse monoclonal antibody that detects TKS5 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

SH3 and PX domain-containing protein 2A (UniProt: Q5TCZ1; also known as Adapter protein TKS5, Five SH3 domain-containing protein, SH3 multiple domains protein 1, Tyrosine kinase substrate with five SH3 domains) is encoded by the SH3PXD2A (also known as FISH, KIAA0418, SH3MD1, TKS5) gene (Gene ID: 9644) in human. Adapter protein TKS5 is a scaffolding protein that is required for podosome formation, degradation of the extracellular matrix, and for invasion in some cancer cells. Three isoforms of TKS5 have been described that are produced by alternative splicing. TKS5 is a cytoplasmic protein in normal cells, but it localizes to podosomes in SRC-transformed cells. It is found in several cancer cell lines, particularly invasive breast carcinomas and melanomas. TKS5 contains one PX (phx homology) domain and five SH3 domains. The PX domain is required for podosome localization because of its ability to bind phosphoinosiltols, such as PtdIns3P and PtdIns(3,4)P2 and to lesser extent PtdIns5P and PtdIns(3,5)P2. Its binding with ADAM12, ADAM15 and ADAM19 is mediated by the fifth SH3 domain. The intramolecular interaction of the PX domain with the third SH3 domain maintains the protein in the cytoplasm and phosphorylation disrupts this interaction, resulting in the redistribution of the protein from cytoplasm to the perimembrane region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TKS5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 13H6.3
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TKS5 in human breast cancer tissue.
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Adapter protein TKS5;Five SH3 domain-containing protein;SH3 multiple domains protein 1;Tyrosine kinase substrate with five SH3 domains;SH3 and PX domain-containing protein 2A
Gene Symbols SH3PXD2A;FISH;KIAA0418;SH3MD1;TKS5
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 60 amino acids from the C-terminal half of human SH3 and PX domain-containing protein 2A (Adapter protein TKS5).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone 13H6.3 detects SH3 and PX domain-containing protein 2A (adapter protein TKS5) in human cells. It targets and epitope with in 60 amino acids crom the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.