Learn More
TKT Mouse anti-Human, Clone: OTI5H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500892

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Transketolase encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
TKT | |
Monoclonal | |
Unconjugated | |
TKT | |
CFLL4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7086 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI5H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P29401 | |
TKT | |
Full length human recombit protein of human TKT produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.