Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TKTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544093 Supplier Invitrogen™ Supplier No. PA544093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QIQTSRNLDP QPPIEDSPEV NITDVRMTSP PDYRVGDKIA TRKACGLALA Sequence homology: Dog: 83%; Guinea Pig: 79%; Horse: 77%; Human: 100%; Mouse: 85%; Rabbit: 77%; Rat: 77%.

Strong TKTL1 protein expression has been correlated with a certain type of glucose metabolism (aerobic glycolysis; Warburg effect) and to cells which are affected by chronic complications of diabetic patients. In colon and urothelial carcinomas, expression at the protein level is correlated with invasiveness of tumors and poor patient survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TKTL1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TKTL1
Gene Accession No. P51854
Gene Alias C79967; TK 2; TKR; TKT2; tktl1; Transketolase 2; transketolase like 1; transketolase-2; transketolase-like 1; transketolase-like protein 1; Transketolase-related protein
Gene Symbols TKTL1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TKTL1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8277
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.