Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TKTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551394 Supplier Invitrogen™ Supplier No. PA551394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFSHPTSCSS SSEIMSVLFF YIMRYKQSDP ENPDNDRFVL AKRLSFVDVA TGWLGQGLGV ACGMAYTGKY FDRASYRVFC LMSDGESS Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 73%.

Strong TKTL1 protein expression has been correlated with a certain type of glucose metabolism (aerobic glycolysis; Warburg effect) and to cells which are affected by chronic complications of diabetic patients. In colon and urothelial carcinomas, expression at the protein level is correlated with invasiveness of tumors and poor patient survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TKTL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TKTL1
Gene Accession No. P51854
Gene Alias C79967; TK 2; TKR; TKT2; tktl1; Transketolase 2; transketolase like 1; transketolase-2; transketolase-like 1; transketolase-like protein 1; Transketolase-related protein
Gene Symbols TKTL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TKTL1. Recombinant protein control fragment (Product #RP-91979).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.