Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TKTL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531192 Supplier Invitrogen™ Supplier No. PA531192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, Jurkat, Raji. Predicted reactivity: Mouse (80%), Rat (83%), Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Plays an essential role in total transketolase activity and cell proliferation in cancer cells; after transfection with anti-TKTL1 siRNA, total transketolase activity dramatically decreases and proliferation was significantly inhibited in cancer cells. Plays a pivotal role in carcinogenesis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TKTL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TKTL2
Gene Accession No. Q9H0I9
Gene Alias 4933401I19Rik; RGD1304767; testis tissue sperm-binding protein Li 39a; TKTL2; transketolase like 2; transketolase-like 2; transketolase-like protein 2
Gene Symbols TKTL2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human TKTL2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.