Learn More
Description
PROTAC comprised of a highly promiscuous kinase inhibitor conjugated to the cereblon E3 ubiquitin ligase ligand, pomalidomide (Cat. No. 6302). Degrades a range of kinases in MOLT-4 and MOLM-14 cell lines.
Negative control TL 13-27 (Cat. No. 6525) is also available.
Negative control TL 13-27 (Cat. No. 6525) is also available.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-(2-(2-(2-(4-(4-((5-Chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)phenyl)piperazin-1-yl)ethoxy)ethoxy)ethyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)acetamide |
| CAS | 2250025-88-6 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Molecular Formula | C44H51ClN10O9S |
| Formula Weight | 931.46 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

