Learn More
Description
Selective anaplastic lymphoma kinase (ALK) degrader (DC50 values are 10 and 180 nM in H3122 and Karpas 299 cells, respectively). Comprises the cereblon E3 ligase ligand Pomalidomide (Cat. No. 6302), conjugated to an ALK inhibitor. Inhibits proliferation of ALK-positive cancer cell lines. Exhibits higher selectivity for ALK over Aurora A kinase compared with TL 13 -112 (Cat.No. 6745). Maximum degradation is exhibited at 16 h.
Negative control TL 13-22 (Cat. No. 6747) also available.
Negative control TL 13-22 (Cat. No. 6747) also available.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-(2-(2-(2-(4-(4-((5-Chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)-3-methoxyphenyl)piperazin-1-yl)ethoxy)ethoxy)ethyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)acetamide |
| CAS | 2229037-04-9 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Molecular Formula | C45H53ClN10O10S |
| Formula Weight | 961.48 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

