Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TL132 Monoclonal Antibody (3A2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523186
Description
Peptide Sequence: HIKPIYNLYA ISCHSGILSG GHYVTYAKNP NCKWYCYNGS ICEEHHPDEI DTDSAYILFY EQQRIDYAQF LPKIDGKKMA DTSSMDEDFE SDYEKYCVLQ.

Specifications
TL132 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
0 | |
USP32P2 | |
LOC220594 (NP_665808, 255 a.a. ~ 354 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot, Immunocytochemistry | |
3A2 | |
Unconjugated | |
USP32P2 | |
TL132; ubiquitin specific peptidase 32 pseudogene 2; USP32P2 | |
Mouse | |
Affinity chromatography | |
RUO | |
220594 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction