Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech TL1A Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P24050UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P24050UG 50 μg
500P240100 100 μg
500P2401MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P24050UG Supplier PeproTech Supplier No. 500P24050UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: QLRAQGEASV QFQALKGQEF APSHQQVYAP LRADGDKPRA HLTVVRQTPT QHFKNQFPAL HWEHELGLAF TKNRMNYTNK FLLIPESGDY FIYSQVTFRG MTSECSEIRQ AGRPNKPDSI TVVITKVTDS YPEPTQLLMG TKSVCEVGSN WFQPIYLGAM FSLQEGDKLM VNVSDISLVD YTKEDKTFFG AFLL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TL-1A. Anti-Human TL-1A-specific antibody was purified by affinity chromatography employing an immobilized Human TL-1A matrix. Sandwich ELISA: To detect Human TL-1A by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TL-1A (500-P240Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human TL-1A. Western Blot: To detect Human TL-1A by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human TL-1A is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This protein is abundantly expressed in endothelial cells, but is not expressed in either B or T cells. The expression of this protein is inducible by TNF and IL-1 alpha. This cytokine is a ligand for receptor TNFRSF25 and decoy receptor TNFRSF21/DR6. It can activate NF-kappaB and MAP kinases, and acts as an autocrine factor to induce apoptosis in endothelial cells. This cytokine is also found to inhibit endothelial cell proliferation, and thus may function as an angiogenesis inhibitor. An additional isoform encoded by an alternatively spliced transcript variant has been reported but the sequence of this transcript has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TL1A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TNFSF15
Gene Accession No. O95150
Gene Alias bM20K13.3 (tumor necrosis factor (ligand) superfamily, member 15); MGC129934; MGC129935; TL1; TL1A; TNF ligand-related molecule 1; TNF superfamily ligand TL1A; TNFSF15; TNLG1B; tumor necrosis factor (ligand) superfamily member 15; tumor necrosis factor (ligand) superfamily, member 15; tumor necrosis factor ligand 1B; tumor necrosis factor ligand superfamily member 15; Tumor necrosis factor ligand superfamily member 15, membrane form; Tumor necrosis factor ligand superfamily member 15, secreted form; tumor necrosis factor superfamily member 15; vascular endothelial cell growth inhibitor; vascular endothelial growth inhibitor; vascular endothelial growth inhibitor-192A; Vegi; VEGI192A
Gene Symbols TNFSF15
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human TL-1A.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9966
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.