Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TL1A/TNFSF15 Antibody (2116C) - Azide and BSA Free, R&D Systems™
SDP

Rabbit Monoclonal Antibody

$243.50 - $560.50

Specifications

Antigen TL1A/TNFSF15
Clone 2116C
Concentration 1.0 mg/mL
Dilution Western Blot, Blocking/Neutralizing
Applications Western Blot, Blocking Assay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB7442SP
SDP
View Documents
Novus Biologicals
MAB7442SP
25 μg
Each for $243.50
Only null left
Add to Cart
 
MAB7442
SDP
View Documents
Novus Biologicals
MAB7442
100 μg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Western Blot, Block/Neutralize.
Specifications

Specifications

TL1A/TNFSF15
1.0 mg/mL
Western Blot, Blocking Assay
Unconjugated
Rabbit
Apoptosis, Biologically Active Proteins, Cell Cycle and Replication
PBS with No Preservative
9966
Partial recombinant human TL1A protein (amino acids 72-251) [UniProt O95150]
Primary
Store at -20C. Avoid freeze-thaw cycles.
2116C
Western Blot, Blocking/Neutralizing
Monoclonal
Purified
RUO
Human
MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1
TNFSF15
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.