Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TL1A/TNFSF15 Antibody (2116C), DyLight 755, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB7442IR
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Western Blot.Specifications
TL1A/TNFSF15 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TNFSF15 | |
Partial recombinant human TL1A protein (amino acids 72-251) [UniProt O95150] | |
100 μL | |
Apoptosis, Cell Cycle and Replication | |
9966 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
2116C | |
Western Blot | |
MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction