Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TL1A/TNFSF15 Antibody (TAN2-2), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281120G
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
TL1A/TNFSF15 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
TNFSF15 | |
This antibody was raised by immunising rats with a soluble recombinant TL1A/TNFSF15 (T77-L252):Fc protein. | |
0.1 mL | |
Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
9966 | |
Mouse | |
Purified |
Flow Cytometry, Immunofluorescence | |
TAN2-2 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody is specific for TL1A/TNFSF15, a member of the TNF ligand family. TL1A/TNFSF15 is expressed by endothelial cells, tissue macrophages, monocyte-derived dendritic cells, plasma cells and lamina propria-derived lymphocytes. Among peripheral blood cells, TL1A/TNFSF15 has been shown to be expressed predominantly by CD4+CCR9+ lymphocytes. Its expression has been shown to be induced by TNF alpha, IL-1 alpha and IFN gamma. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction