Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TL1A/TNFSF15 Antibody (VEGI/2052R), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308802B
Description
TL1A/TNFSF15 Monoclonal antibody specifically detects TL1A/TNFSF15 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
TL1A/TNFSF15 | |
Monoclonal | |
Biotin | |
MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
Recombinant full-length human TL1A/TNFSF15 protein (Uniprot: O95150) | |
100 μg | |
Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
9966 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin), Protein Array | |
VEGI/2052R | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction