Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TL4-12
GREENER_CHOICE

Catalog No. 591010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-101-0 10 mg
59-105-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-101-0 Supplier Tocris Bioscience™ Supplier No. 5910/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent MAP4K2 (GCK) inhibitor

TL4-12 is a potent MAP4K2 (GCK) inhibitor (IC50 = 37 nM). Exhibits >70-fold selectivity for MAP4K2 over TAK1 (IC50 = 2.7 μM for TAK1). Inhibits IL-1 and TGFβ-induced p38 MAPK phosphorylation, in vitro.

Specifications

Chemical Name or Material 4-Methyl-3-[[6-(methylamino)-4-pyrimidinyl]oxy]-N-[3-(4-methyl-1-piperazinyl)-5-(trifluoromethyl)phenyl]benzamide
CAS 1620820-12-3
Quantity 10 mg
Molecular Formula C25H27F3N6O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.