Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLCD1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548778
Description
Calcium channel facilitator that increases calcium flux by generating a larger window current and slowing inactivation of the L-type CACNA1C/CaV1.2 channel. Regulation of intracellular calcium by Calfacilitin is required for neural plate formation.
Specifications
TLCD1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96CP7 | |
TLCD1 | |
KLH conjugated synthetic peptide between 218-247 amino acids from the C-terminal region of human TLCD1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TLCD1 | |
Calfacilitin, TLC domain-containing protein 1, TLCD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116238 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction