Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLDC1 Antibody (OTI5D9), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272091C
Description
TLDC1 Monoclonal specifically detects TLDC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TLDC1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MEAK7 | |
Full length recombinant protein of human KIAA1609 (NP_065998) produced in HEK293T cells. | |
0.1 mL | |
57707.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5D9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
hypothetical protein LOC57707, KIAA1609, MGC25024 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction