Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLE Pan Polyclonal Antibody
TLE Pan Polyclonal Antibody
Supplier: Thermo Scientific PA517241
Description
TLE Pan Polyclonal Antibody for Western Blot

Specifications
TLE Pan | |
Polyclonal | |
Unconjugated | |
tle2 | |
2610103N05Rik; 5730411M05Rik; AA792082; B lymphocyte gene 1; BCE1; Bce-1; C230057C06Rik; E(Sp1) homolog; E(spI); enhancer of split groucho; enhancer of split groucho 3; enhancer of split groucho 4; enhancer of split groucho-like protein 1; Enhancer of split groucho-like protein 2; enhancer of split groucho-like protein 3; Esg; esg1; ESG-1; ESG2; ESG-2; esg3; ESG-3; ESG4; ESG-4; Esp2; Esp3; ESTM13; Estm14; fc18h03; GRG1; Grg-1; GRG2; GRG3; Grg-3; Grg3a; Grg3b; GRG4; Grg-4; gro; groucho protein; groucho-related gene 4; groucho-related protein; groucho-related protein 1; Groucho-related protein 4; HsT18976; KIAA1261; KIAA1547; mKIAA1547; mKIAA4188; Protein ESP2; rTLE3; sb:eu1034; sb:eu408; si:ch211-81i17.1; si:dkey-42p8.3; TLE family member 2, transcriptional corepressor a; TLE family member 2, transcriptional corepressor b; tle1; tle1.S; Tle2; tle2.L; tle2a; tle2b; TLE3; tle3a; tle3b; tle4; tle4.S; Tle4l; transducin like enhancer of split 1; transducin like enhancer of split 1 S homeolog; transducin like enhancer of split 2; transducin like enhancer of split 2 L homeolog; transducin like enhancer of split 2a; transducin like enhancer of split 2b; transducin like enhancer of split 3; transducin like enhancer of split 4; transducin like enhancer of split 4 S homeolog; transducin-like enhancer of split 1 (E(sp1) homolog); transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 1, homolog of Drosophila E(spl); transducin-like enhancer of split 2; transducin-like enhancer of split 2 (E(sp1) homolog); transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2 homolog; transducin-like enhancer of split 2, homolog of Drosophila E(sp1); transducin-like enhancer of split 2, homolog of Drosophila E(spl); transducin-like enhancer of split 2; transducin-like enhancer protein 2a; transducin-like enhancer of split 3; transducin-like enhancer of split 3 (E(sp1) homolog) a; transducin-like enhancer of split 3 (E(sp1) homolog) b; transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 3 homolog of Drosophila; transducin-like enhancer of split 3 short isoform; transducin-like enhancer of split 3, E(spl) homolog; transducin-like enhancer of split 3, homolog of Drosophila E(sp1); transducin-like enhancer of split 3, homolog of Drosophila E(spl); transducin-like enhancer of split 4; transducin-like enhancer of split 4 (E(sp1) homolog); transducin-like enhancer of split 4 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 4 (homolog of Drosophila E(spl)) - like; transducin-like enhancer of split 4, E(spl) homolog; transducin-like enhancer of split 4, homolog of Drosophila E(sp1); transducin-like enhancer of split 4, homolog of Drosophila E(spl); transducin-like enhancer protein 1; Transducin-like enhancer protein 2; transducin-like enhancer protein 3; Transducin-like enhancer protein 4; wu:fb96a10; wu:fc18h03; X83332; X83333; XELAEV_18001139mg; XELAEV_18006642mg; XELAEV_18010139mg; Xesg1; xgrg1; xgrg3; Xgrg4; XGrg-4; X-groucho; XTCF-4; Xtle4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21885, 21886, 21887, 21888, 7088, 7089, 7090, 7091 | |
-20°C | |
Liquid |
Western Blot | |
261 μg/mL | |
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
Q04724, Q04725, Q04726, Q04727, Q08122, Q62440, Q62441, Q9WVB2 | |
Tle1, tle2, TLE3, TLE4 | |
Synthetic peptide corresponding to residues surrounding Asp760 of human TLE1. | |
100 μL | |
Primary | |
Human, Mouse, Monkey, Drosophila, Rat, X. laevis, Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction