Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TLE1 Recombinant Rabbit Monoclonal Antibody (6Q5X1)
GREENER_CHOICE

Catalog No. PIMA535377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535377 Supplier Invitrogen™ Supplier No. MA535377
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: LQPPGIPPLG GSAGLLALSS ALSGQSHLAI KDDKKHHDAE HHRDREPGTS NSLLVPDSLR GTDKRRNGPE FSNDIKKRKV DDKDSSHYDS DGDKSDDNLV V.

Transcriptional corepressor that binds to a number of transcription factors. Inhibits NF-kappa-B-regulated gene expression. Inhibits the transcriptional activation mediated by FOXA2, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with dominant-negative AES. Unusual function as coactivator for ESRRG.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLE1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6Q5X1
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TLE1
Gene Accession No. Q04724, Q62440
Gene Alias C230057C06Rik; E(Sp1) homolog; enhancer of split groucho; enhancer of split groucho-like protein 1; esg; ESG1; Estm14; grg1; Grg-1; Groucho-related protein 1; RP11-203D19.1; TLE 1; TLE1; tle1.S; tle3b; Tle4l; transducin like enhancer of split 1; transducin like enhancer of split 1 S homeolog; transducin-like enhancer of split 1 (E(sp1) homolog); transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 1, homolog of Drosophila E(spl); transducin-like enhancer of split 3 (E(sp1) homolog) b; transducin-like enhancer of split 4 (homolog of Drosophila E(spl)) - like; transducin-like enhancer protein 1; XELAEV_18001139mg; xgrg1
Gene Symbols TLE1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 150-250 of human TLE1 (Q04724).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21885, 362533, 7088
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.