Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE1 Mouse anti-Human, Biotin, Clone: OTI1F9, liquid, TrueMAB™
SDP

Catalog No. 501677800
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7800 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7800 Supplier OriGene Supplier No. TA700293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcriptional corepressor that binds to a number of transcription factors. Inhibits NF-kappa-B-regulated gene expression. Inhibits the transcriptional activation mediated by FOXA2, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with domit-negative AES. Unusual function as coactivator for ESRRG.

Specifications

Antigen TLE1
Applications ELISA
Classification Monoclonal
Clone OTI1F9
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide
Gene TLE1
Gene Accession No. Q04724
Gene Alias ESG, ESG1, GRG1
Gene Symbols TLE1
Host Species Mouse
Immunogen Full length human recombit protein of human TLE1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7088
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.