Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TLE1 Rabbit anti-Human, Unconjugated, Clone: TLE1/2946R, Abnova™

Catalog No. 89339659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-659 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-659 Supplier Abnova Corporation Supplier No. RAB00644
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

TLE1 Monoclonal antibody specifically detects TLE1 in Human samples. It is validated for IHC-P.

Specifications

Antigen TLE1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TLE1/2946R
Conjugate Unconjugated
Description transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila)
Gene TLE1
Gene Alias ESG, ESG1, GRG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 175-338 of human TLE1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7088
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.